Tisk…

1-Bromo-1H-pyrazol-5-amine 95%

Dodavatel: Apollo Scientific

OR947550-1G OR947550-5G
APOSOR947550-1GEA 0 CZK
APOSOR947550-1G APOSOR947550-5G
1-Bromo-1H-pyrazol-5-amine 95%
1-Bromo-1H-pyrazol-5-amine
Vzorec: C₃H₄BrN₃
MW: 161,990 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26142695
CAS číslo: 1823380-61-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR