Tisk…

(2-Oxo-2,3-dihydro-1H-indol-3-yl)acetic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(2-Oxoindolin-3-yl)acetic acid

OR947497-1G
APOSOR947497-1GEA 4070 CZK
APOSOR947497-1G
(2-Oxo-2,3-dihydro-1H-indol-3-yl)acetic acid 95%
(2-Oxo-2,3-dihydro-1H-indol-3-yl)acetic acid
Vzorec: C₁₀H₉NO₃
MW: 191,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09035909
CAS číslo: 2971-31-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR