4-Methoxy-3-methylphenylacetonitrile 95%
Dodavatel: Apollo Scientific
OR947347-1G
OR947347-5G
APOSOR947347-1GEA
6020
CZK
APOSOR947347-1G
APOSOR947347-5G
4-Methoxy-3-methylphenylacetonitrile 95%
4-Methoxy-3-methylphenylacetonitrile
Vzorec:
C₁₀H₁₁NO Storage Temperature: Ambient |
MDL Number:
MFCD00040891 CAS číslo: 75391-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...