4-Acetoxymethylbenzeneboronic acid 97%
Dodavatel: Apollo Scientific
OR9468-1G
OR9468-5G
APOSOR9468-1GEA
3940
CZK
APOSOR9468-1G
APOSOR9468-5G
4-Acetoxymethylbenzeneboronic acid 97%
4-Acetoxymethylbenzeneboronic acid
Vzorec:
C₉H₁₁BO₄ Bod tání: 81…83 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08235082 CAS číslo: 326496-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...