Tisk…

3-Cyano-N-methylbenzamide 98%

Dodavatel: Apollo Scientific

OR946459-1G OR946459-5G
APOSOR946459-1GEA 4400 CZK
APOSOR946459-1G APOSOR946459-5G
3-Cyano-N-methylbenzamide 98%
3-Cyano-N-methylbenzamide
Vzorec: C₉H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD09939344
CAS číslo: 363186-09-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR