3-(2-Methoxyethoxy)benzaldehyde 98%
Dodavatel: Apollo Scientific
OR946448-1G
OR946448-250MG
APOSOR946448-1GEA
10350
CZK
APOSOR946448-1G
APOSOR946448-250MG
3-(2-Methoxyethoxy)benzaldehyde 98%
3-(2-Methoxyethoxy)benzaldehyde
Vzorec:
C₁₀H₁₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD08144122 CAS číslo: 73244-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...