Tisk…

1-Butyl-5-oxopyrrolidine-3-carboxylic acid 96%

Dodavatel: Apollo Scientific

OR946386-1G OR946386-5G
APOSOR946386-1GEA 8270 CZK
APOSOR946386-1G APOSOR946386-5G
1-Butyl-5-oxopyrrolidine-3-carboxylic acid 96%
1-Butyl-5-oxopyrrolidine-3-carboxylic acid
Vzorec: C₉H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD04989720
CAS číslo: 43094-86-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR