Tisk…

S-Butyl thioacetate 95%

Dodavatel: Apollo Scientific

Synonyms: S-n-Butylthioacetate

OR945643-5G
APOSOR945643-5GEA 6450 CZK
APOSOR945643-5G
S-Butyl thioacetate 95%
S-Butyl thioacetate
Vzorec: C₆H₁₂OS
MW: 132,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039956
CAS číslo: 928-47-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR