4-Chloro-2-methyl-6-propylpyrimidine 95%
Dodavatel: Apollo Scientific
OR944950-1G
OR944950-5G
APOSOR944950-1GEA
4770
CZK
APOSOR944950-1G
APOSOR944950-5G
4-Chloro-2-methyl-6-propylpyrimidine 95%
4-Chloro-2-methyl-6-propylpyrimidine
Vzorec:
C₈H₁₁ClN₂ MW: 170.64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07365353 CAS číslo: 89967-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...