Tisk…

5-Formyl-2-methylbenzonitrile 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR944886-1G OR944886-5G
APOSOR944886-1GEA 8300 CZK
APOSOR944886-1G APOSOR944886-5G
5-Formyl-2-methylbenzonitrile 97%
5-Formyl-2-methylbenzonitrile
Vzorec: C₉H₇NO
Storage Temperature: Ambient
MDL Number: MFCD12197728
CAS číslo: 27613-36-1
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR