Tisk…

2,5-Dichloroisonicotinic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,5-Dichloropyridine-4-carboxylic acid

OR9438-100G OR9438-25G
APOSOR9438-100GEA 7410 CZK
APOSOR9438-100G APOSOR9438-25G
2,5-Dichloroisonicotinic acid
2,5-Dichloroisonicotinic acid
Vzorec: C₆H₃Cl₂NO₂
MW: 192 g/mol
Bod tání: 235…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00466640
CAS číslo: 88912-26-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR