2-Methoxyquinoline-3-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR943652-1G
APOSOR943652-1GEA
2200
CZK
APOSOR943652-1G
2-Methoxyquinoline-3-carbaldehyde 95%
2-Methoxyquinoline-3-carbaldehyde
Vzorec:
C₁₁H₉NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD03864951 CAS číslo: 139549-06-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...