1-Chloro-2-(ethylsulphonyl)ethane 95%
Dodavatel: Apollo Scientific
OR943614-1G
OR943614-250MG
APOSOR943614-1GEA
13860
CZK
APOSOR943614-1G
APOSOR943614-250MG
1-Chloro-2-(ethylsulphonyl)ethane 95%
1-Chloro-2-(ethylsulphonyl)ethane
Vzorec:
C₄H₉ClO₂S MW: 156,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01752806 CAS číslo: 25027-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...