Tisk…

4-Hydroxybenzimidazole 95%

Dodavatel: Apollo Scientific

Synonyms: 1H-benzo[d]imidazol-4-ol, 1H-Benzimidazol-4-ol

OR943594-1G OR943594-250MG
APOSOR943594-1GEA 2570 CZK
APOSOR943594-1G APOSOR943594-250MG
4-Hydroxybenzimidazole 95%
4-Hydroxybenzimidazole
Vzorec: C₇H₆N₂O
MW: 134,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00963978
CAS číslo: 67021-83-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR