2-Chloro-N-cyclopentylacetamide 95%
Dodavatel: Apollo Scientific
OR943567-1G
OR943567-250MG
OR943567-5G
APOSOR943567-1GEA
3870
CZK
APOSOR943567-1G
APOSOR943567-250MG
APOSOR943567-5G
2-Chloro-N-cyclopentylacetamide 95%
2-Chloro-N-cyclopentylacetamide
Vzorec:
C₇H₁₂ClNO Storage Temperature: Ambient |
MDL Number:
MFCD00297016 CAS číslo: 125674-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...