Tisk…

2,4-Dimethylbenzylamine 98%

Dodavatel: Apollo Scientific

OR943193-1G
APOSOR943193-1GEA 3150 CZK
APOSOR943193-1G
2,4-Dimethylbenzylamine 98%
2,4-Dimethylbenzylamine
Vzorec: C₉H₁₃N
MW: 135,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00025575
CAS číslo: 94-98-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR