2-(1H-Pyrrolo[2,3-b]pyridin-1-yl)acetic acid 95%
Dodavatel: Apollo Scientific
OR943139-1G
OR943139-500MG
APOSOR943139-1GEA
21490
CZK
APOSOR943139-1G
APOSOR943139-500MG
2-(1H-Pyrrolo[2,3-b]pyridin-1-yl)acetic acid 95%
2-(1H-Pyrrolo[2,3-b]pyridin-1-yl)acetic acid
Vzorec:
C₉H₈N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09834895 CAS číslo: 1048913-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...