Tisk…

(3-Phenyl-1H-pyrazol-1-yl)acetic acid

Dodavatel: Apollo Scientific
Varování

OR9431-1G OR9431-250MG OR9431-100MG
APOSOR9431-1GEA 12530 CZK
APOSOR9431-1G APOSOR9431-250MG APOSOR9431-100MG
(3-Phenyl-1H-pyrazol-1-yl)acetic acid
(3-Phenyl-1H-pyrazol-1-yl)acetic acid
Vzorec: C₁₁H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00665957
CAS číslo: 959574-98-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR