Tisk…

4-Pyridazineacetonitrile

Dodavatel: Apollo Scientific

OR943016-1G OR943016-250MG OR943016-500MG
APOSOR943016-1GEA 48600 CZK
APOSOR943016-1G APOSOR943016-250MG APOSOR943016-500MG
4-Pyridazineacetonitrile
4-Pyridazineacetonitrile
Vzorec: C₆H₅N₃
Storage Temperature: Ambient
MDL Number: MFCD17015867
CAS číslo: 1142927-95-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR