Tisk…

5-Bromo-N-methylpyrazin-2-amine 95%

Dodavatel: Apollo Scientific

OR942676-1G OR942676-250MG
APOSOR942676-1GEA 17990 CZK
APOSOR942676-1G APOSOR942676-250MG
5-Bromo-N-methylpyrazin-2-amine 95%
5-Bromo-N-methylpyrazin-2-amine
Vzorec: C₅H₆BrN₃
MW: 188,03 g/mol
Storage Temperature: Ambient
CAS číslo: 446286-92-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR