Tisk…

6-tert-Butyl-o-cresol 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(tert-Butyl)-6-methylphenol, 2-tert-Butyl-6-methylphenol

OR942617-25G OR942617-100G OR942617-500G
APOSOR942617-25GEA 3740 CZK
APOSOR942617-25G APOSOR942617-100G APOSOR942617-500G
6-tert-Butyl-o-cresol 95%
6-tert-Butyl-o-cresol
Vzorec: C₁₁H₁₆O
MW: 164,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002239
CAS číslo: 2219-82-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR