Dimethyl methylsuccinate 95%
Dodavatel: Apollo Scientific
Synonyms:
1,4-dimethyl (2S)-2-methylbutanedioate, Dimethyl (2S)-2-methylsuccinate, (S)-(-)-Methylsuccinic acid dimethyl ester
OR942307-5G
APOSOR942307-5GEA
1950
CZK
APOSOR942307-5G
Dimethyl methylsuccinate 95%
Dimethyl methylsuccinate
Vzorec:
C₇H₁₂O₄ MW: 160,168 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008449 CAS číslo: 1604-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...