Tisk…

2-Vinylphenyl acetate 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Acetoxystyrene, Acetic Acid 2-Vinylphenyl Ester

OR942180-1G OR942180-250MG
APOSOR942180-1GEA 2730 CZK
APOSOR942180-1G APOSOR942180-250MG
2-Vinylphenyl acetate 95%
2-Vinylphenyl acetate
Vzorec: C₁₀H₁₀O₂
MW: 162.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093430
CAS číslo: 63600-35-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR