Tisk…

3-[Ethyl(methyl)carbamoyl]benzeneboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-(Ethyl(methyl)carbamoyl)phenyl)boronic acid, 3-[(N-Ethyl-N-methylamino)carbonyl]benzeneboronic acid

OR9416-1G OR9416-5G
APOSOR9416-1GEA 4770 CZK
APOSOR9416-1G APOSOR9416-5G
3-[Ethyl(methyl)carbamoyl]benzeneboronic acid 98%
3-[Ethyl(methyl)carbamoyl]benzeneboronic acid
Vzorec: C₁₀H₁₄BNO₃
MW: 207,04 g/mol
Bod tání: 73…76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08056369
CAS číslo: 871333-07-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR