5-Methoxybenzofuran-2-carboxylic acid 95%
Dodavatel: Apollo Scientific
OR941323-1G
OR941323-5G
APOSOR941323-1GEA
1780
CZK
APOSOR941323-1G
APOSOR941323-5G
5-Methoxybenzofuran-2-carboxylic acid 95%
5-Methoxybenzofuran-2-carboxylic acid
Vzorec:
C₁₀H₈O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00079771 CAS číslo: 10242-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...