Tisk…

4-tert-Butyl-o-cresol 95%

Dodavatel: Apollo Scientific

Synonyms: 4-tert-Butyl-2-methylphenol, 4-(tert-Butyl)-2-methylphenol

OR941143-10G OR941143-1G OR941143-5G
APOSOR941143-10GEA 12830 CZK
APOSOR941143-10G APOSOR941143-1G APOSOR941143-5G
4-tert-Butyl-o-cresol 95%
4-tert-Butyl-o-cresol
Vzorec: C₁₁H₁₆O
MW: 164,2400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00020047
CAS číslo: 98-27-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR