2-(4-Bromophenyl)-2-methylpropanenitrile 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR9411-1G
OR9411-250MG
APOSOR9411-1GEA
787
CZK
APOSOR9411-1G
APOSOR9411-250MG
2-(4-Bromophenyl)-2-methylpropanenitrile 98%
2-(4-Bromophenyl)-2-methylpropanenitrile
Vzorec:
C₁₀H₁₀BrN Storage Temperature: Ambient |
MDL Number:
MFCD02684219 CAS číslo: 101184-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...