Tisk…
Ethylene glycol mono-tert-butyl ether 95%
Dodavatel: Apollo Scientific
Synonyms: Ethyleneglycolmono-tert-butylether, tert-Butyl Cellosolve, 2-tert-Butoxyethanol
OR940810-100G OR940810-1KG OR940810-500G
APOSOR940810-100GEA 3540 CZK
APOSOR940810-100G APOSOR940810-1KG APOSOR940810-500G
Ethylene glycol mono-tert-butyl ether 95%
Ethylene glycol mono-tert-butyl ether
Vzorec: C₆H₁₄O₂
MW: 118.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059004
CAS číslo: 7580-85-0