3-Methoxybutyl acetate 95%
Dodavatel: Apollo Scientific
Synonyms:
Acetic Acid 3-Methoxybutyl Ester
OR940548-100G
OR940548-1KG
OR940548-500G
APOSOR940548-100GEA
3540
CZK
APOSOR940548-100G
APOSOR940548-1KG
APOSOR940548-500G
3-Methoxybutyl acetate 95%
3-Methoxybutyl acetate
Vzorec:
C₇H₁₄O₃ MW: 146.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00043928 CAS číslo: 4435-53-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...