Tisk…

sec-Butyl butyrate 95%

Dodavatel: Apollo Scientific

Synonyms: Butyric Acid sec-Butyl Ester

OR939965-10G OR939965-1G OR939965-5G
APOSOR939965-10GEA 9330 CZK
APOSOR939965-10G APOSOR939965-1G APOSOR939965-5G
sec-Butyl butyrate 95%
sec-Butyl butyrate
Vzorec: C₈H₁₆O₂
MW: 144.21 g/mol
Storage Temperature: Ambient
CAS číslo: 819-97-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR