1-Thien-2-ylpropan-1-amine 95%
Dodavatel: Apollo Scientific
Synonyms:
1-(Thiophen-2-yl)propan-1-amine hydrochloride
OR939957-1G
OR939957-250MG
OR939957-500MG
APOSOR939957-1GEA
9530
CZK
APOSOR939957-1G
APOSOR939957-250MG
APOSOR939957-500MG
1-Thien-2-ylpropan-1-amine 95%
1-Thien-2-ylpropan-1-amine
Vzorec:
C₇H₁₁NS MW: 141,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09040987 CAS číslo: 6315-55-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...