Tisk…

Ethyl dichloroacetate 98%

Dodavatel: Apollo Scientific

OR939883-100G OR939883-25G
APOSOR939883-100GEA 9100 CZK
APOSOR939883-100G APOSOR939883-25G
Ethyl dichloroacetate 98%
Ethyl dichloroacetate
Vzorec: C₄H₆Cl₂O₂
MW: 157 g/mol
Storage Temperature: Ambient
CAS číslo: 535-15-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR