(+)-Limonene 1,2-epoxide 95%
Dodavatel: Apollo Scientific
Synonyms:
(4R)-1,2-Epoxy-p-menth-8-ene
OR939871-100G
OR939871-25G
OR939871-10G
APOSOR939871-100GEA
11410
CZK
APOSOR939871-100G
APOSOR939871-25G
APOSOR939871-10G
(+)-Limonene 1,2-epoxide 95%
(+)-Limonene 1,2-epoxide
MW:
152,23 g/mol Storage Temperature: Refrigerator |
CAS číslo:
203719-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...