Tisk…

Imidazo[1,5-a]pyridine-3-carboxaldehyde 98%

Dodavatel: Apollo Scientific

Synonyms: Imidazo[1,5-a]pyridine-3-carbaldehyde, 3-Formylimidazo[1,5-a]pyridine

OR939831-1G OR939831-5G
APOSOR939831-1GEA 2480 CZK
APOSOR939831-1G APOSOR939831-5G
Imidazo[1,5-a]pyridine-3-carboxaldehyde 98%
Imidazo[1,5-a]pyridine-3-carboxaldehyde
Vzorec: C₈H₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD08752619
CAS číslo: 56671-66-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR