Tisk…

1-Ethyl-2-methylindole 95%

Dodavatel: Apollo Scientific

OR939729-10G OR939729-1G OR939729-5G
APOSOR939729-10GEA 8070 CZK
APOSOR939729-10G APOSOR939729-1G APOSOR939729-5G
1-Ethyl-2-methylindole 95%
1-Ethyl-2-methylindole
Vzorec: C₁₁H₁₃N
Storage Temperature: Ambient
MDL Number: MFCD01632168
CAS číslo: 40876-94-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR