Tisk…

Butyl cyanoacetate 95%

Dodavatel: Apollo Scientific

Synonyms: Cyanoacetic acid butyl ester

OR939724-500G
APOSOR939724-500GEA 4070 CZK
APOSOR939724-500G
Butyl cyanoacetate 95%
Butyl cyanoacetate
Vzorec: C₇H₁₁NO₂
MW: 141,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001941
CAS číslo: 5459-58-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR