N,N-Dimethylformamide diisopropyl acetal 95%
Dodavatel: Apollo Scientific
Synonyms:
N,N,N-Trimethyl-1,1-bis(1-methylethoxy)amine, 1,1-Diisopropoxy-N,N-dimethylmethanamine, 1,1-Diisopropoxytrimethylamine
OR939612-25G
OR939612-5G
APOSOR939612-25GEA
5960
CZK
APOSOR939612-25G
APOSOR939612-5G
N,N-Dimethylformamide diisopropyl acetal 95%
N,N-Dimethylformamide diisopropyl acetal
Vzorec:
C₉H₂₁NO₂ MW: 175.27 Bod varu: 79…80 °C Hustota: 0.838 g/cm³ (25 °C) Bod vzplanutí: 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008879 CAS číslo: 18503-89-4 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...