Tisk…

2-Chloro-3-hydroxybenzaldehyde 95%

Dodavatel: Apollo Scientific

OR939587-10G OR939587-25G OR939587-5G
APOSOR939587-10GEA 4770 CZK
APOSOR939587-10G APOSOR939587-25G APOSOR939587-5G
2-Chloro-3-hydroxybenzaldehyde 95%
2-Chloro-3-hydroxybenzaldehyde
Vzorec: C₇H₅ClO₂
MW: 156,57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798118
CAS číslo: 56962-10-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR