Tisk…

(2,3-Dimethoxyphenyl)acetic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(2,3-Dimethoxyphenyl)acetic acid

OR939463-1G OR939463-5G
APOSOR939463-1GEA 2570 CZK
APOSOR939463-1G APOSOR939463-5G
(2,3-Dimethoxyphenyl)acetic acid 95%
(2,3-Dimethoxyphenyl)acetic acid
Vzorec: (C₆H₃(OCH₃)₂)CH₂COOH
MW: 196,2 g/mol
Bod tání: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00060351
CAS číslo: 90-53-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR