Tisk…

2-(3-Methylphenyl)ethanol 95%

Dodavatel: Apollo Scientific

OR939406-1G OR939406-5G
APOSOR939406-1GEA 0 CZK
APOSOR939406-1G APOSOR939406-5G
2-(3-Methylphenyl)ethanol 95%
2-(3-Methylphenyl)ethanol
Vzorec: C₉H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD00002896
CAS číslo: 1875-89-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR