Tisk…

Methyl (trimethylsilyl)acetate 95%

Dodavatel: Apollo Scientific

Synonyms: (Trimethylsilyl)acetic acid methyl ester

OR939403-25G OR939403-5G
APOSOR939403-25GEA 6750 CZK
APOSOR939403-25G APOSOR939403-5G
Methyl (trimethylsilyl)acetate 95%
Methyl (trimethylsilyl)acetate
Vzorec: C₆H₁₄O₂Si
MW: 146.26
Storage Temperature: Ambient
CAS číslo: 2916-76-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR