4-Methylvaleryl chloride 97%
Dodavatel: Apollo Scientific
OR939142-100G
OR939142-25G
OR939142-5G
APOSOR939142-100GEA
29230
CZK
APOSOR939142-100G
APOSOR939142-25G
APOSOR939142-5G
4-Methylvaleryl chloride 97%
4-Methylvaleryl chloride
Vzorec:
C₆H₁₁ClO Storage Temperature: Ambient |
MDL Number:
MFCD00018814 CAS číslo: 38136-29-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...