4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione 95%
Dodavatel: Apollo Scientific
Synonyms:
3,5-Dihydro-4-phenyl-4H-1,2,4-triazole-3,5-dione, PTAD, 4-Phenyl-1,2,4-triazoline-3,5-dione, 4-Phenyl-3H-1,2,4-triazole-3,5(4H)-dione
OR939123-1G
OR939123-5G
APOSOR939123-1GEA
1950
CZK
APOSOR939123-1G
APOSOR939123-5G
4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione 95%
4-Phenyl-3H-1,2,4-triazole-3,5-(4H)-dione
Vzorec:
C₈H₅N₃O₂ MW: 175,1400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003148 CAS číslo: 4233-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...