Tisk…
Dimethyl-trans-4-cyclohexene-1,2-dicarboxylate 95%
Dodavatel: Apollo Scientific
Synonyms: trans-4-Cyclohexene-1,2-dicarboxylic Acid Dimethyl Ester
OR939033-1G OR939033-250MG
APOSOR939033-1GEA 10550 CZK
APOSOR939033-1G APOSOR939033-250MG
Dimethyl-trans-4-cyclohexene-1,2-dicarboxylate 95%
Dimethyl-trans-4-cyclohexene-1,2-dicarboxylate
Vzorec: C₁₀H₁₄O₄
MW: 198,22 g/mol
Storage Temperature: Ambient
CAS číslo: 17673-68-6