2-Isopropoxybenzoic acid 95%
Dodavatel: Apollo Scientific
OR939031-1G
OR939031-250MG
OR939031-500MG
APOSOR939031-1GEA
13860
CZK
APOSOR939031-1G
APOSOR939031-250MG
APOSOR939031-500MG
2-Isopropoxybenzoic acid 95%
2-Isopropoxybenzoic acid
Vzorec:
C₁₀H₁₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00156986 CAS číslo: 63635-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...