Tisk…

2-Amino-5,6-dimethylbenzimidazole 95%

Dodavatel: Apollo Scientific

OR938863-1G OR938863-250MG
APOSOR938863-1GEA 8070 CZK
APOSOR938863-1G APOSOR938863-250MG
2-Amino-5,6-dimethylbenzimidazole 95%
2-Amino-5,6-dimethylbenzimidazole
Vzorec: C₉H₁₁N₃
Bod tání: 220…23 °C (rozkl.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005597
CAS číslo: 29096-75-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR