Tisk…

(1R,2S)-Cyclobutane-1,2-dicarboxylic acid 95%

Dodavatel: Apollo Scientific

Synonyms: cis-Cyclobutane-1,2-dicarboxylic acid

OR938740-1G
APOSOR938740-1GEA 2570 CZK
APOSOR938740-1G
(1R,2S)-Cyclobutane-1,2-dicarboxylic acid 95%
(1R,2S)-Cyclobutane-1,2-dicarboxylic acid
Vzorec: C₆H₈O₄
MW: 144,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019263
CAS číslo: 1461-94-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR