2-Isobutyl-3-methylpyrazine 95%
Dodavatel: Apollo Scientific
Synonyms:
2-Methyl-3-isobutylpyrazine
OR938702-25G
APOSOR938702-25GEA
2030
CZK
APOSOR938702-25G
2-Isobutyl-3-methylpyrazine 95%
2-Isobutyl-3-methylpyrazine
Vzorec:
C₉H₁₄N₂ MW: 150,2209 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00059770 CAS číslo: 13925-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...