Tisk…

trans-4-Ethylcyclohexanol 95%

Dodavatel: Apollo Scientific

OR938558-1G
APOSOR938558-1GEA 5230 CZK
APOSOR938558-1G
trans-4-Ethylcyclohexanol 95%
trans-4-Ethylcyclohexanol
Vzorec: C₈H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD00070694
CAS číslo: 19781-62-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR