Tisk…

2-(Chloromethyl)-1,2-epoxybutane 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(Chloromethyl)-2-ethyloxirane, 2-(Chloromethyl)-1,2-butylene Oxide

OR938507-1G OR938507-250MG OR938507-5G
APOSOR938507-1GEA 5790 CZK
APOSOR938507-1G APOSOR938507-250MG APOSOR938507-5G
2-(Chloromethyl)-1,2-epoxybutane 95%
2-(Chloromethyl)-1,2-epoxybutane
Vzorec: C₅H₉ClO
MW: 120,58 g/mol
Storage Temperature: Ambient
CAS číslo: 75484-32-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR